Learn More
Beckman Coulter Beckman Coulter MLA-150 Fixed-Angle Rotor Package for Optima MAX series ultracentrifuges
Supplier:  Beckman Coulter 393490
Encompass
The MLA-150 Fixed-Angle Rotor Package is a titanium fixed-angle ultracentrifuge rotor designed for high-speed separations in benchtop ultracentrifuges. It accommodates eight 2.0 mL tubes and is intended for applications such as subcellular fractionation, virus pelleting, protein purification, and nucleic acid separations.
  • Constructed from titanium for high strength and corrosion resistance.
  • Rated to 150,000 rpm and up to 989,000 x g for high-speed separations.
  • Holds eight 2.0 mL tubes (total capacity 16 mL).
  • Compact design with a low k-factor for efficient pelleting.
  • Includes floating tube spacers and adapters for secure tube positioning during runs.
Catalog No. NC2827866
Request a Quote
To request a quote for this item, please select Request a Quote to reach our dedicated support team.