Learn More
Beckman Coulter Beckman Coulter MLA-50 Fixed-Angle Aluminum Rotor (A91774) - 6 x 32.4 mL, for Optima MAX-XP ultracentrifuges
Catalog No. NC2831754
Encompass
Beckman Coulter MLA-50 Fixed-Angle Aluminum Rotor (A91774) is a high-speed, six-place fixed-angle rotor engineered for subcellular isolations and preparative ultracentrifugation. It accepts large-volume tubes and is designed for use with compatible benchtop ultracentrifuges.
  • High-speed fixed-angle rotor suitable for subcellular isolations.
  • Holds six 32.4 mL tubes (25 x 77 mm); nominal capacity 194.4 mL.
  • Rated to 50,000 rpm for maximum centrifugal performance.
  • Delivers up to 233,000 x g (max rcf).
  • Constructed from aluminum for strength and durability.
  • Intended for use with compatible benchtop ultracentrifuges; verify instrument compatibility before use.
Catalog No. NC2831754 Supplier Beckman Coulter Supplier No. A91774
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
Beckman Coulter MLA-50 Fixed-Angle Aluminum Rotor (A91774) is a high-speed, six-place fixed-angle rotor engineered for subcellular isolations and preparative ultracentrifugation. It accepts large-volume tubes and is designed for use with compatible benchtop ultracentrifuges.
  • High-speed fixed-angle rotor suitable for subcellular isolations.
  • Holds six 32.4 mL tubes (25 x 77 mm); nominal capacity 194.4 mL.
  • Rated to 50,000 rpm for maximum centrifugal performance.
  • Delivers up to 233,000 x g (max rcf).
  • Constructed from aluminum for strength and durability.
  • Intended for use with compatible benchtop ultracentrifuges; verify instrument compatibility before use.