Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Technologies MLA™ Brand Macro Pipetters
GSA_VAGREENER_CHOICESDP

Catalog No. 21369C Shop All VistaLab Technologies Inc. Products
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
3 mL
4 mL
5 mL
6 mL
8 mL
9 mL
10 mL
Accuracy:
±0.6%
±0.75%
Color:
Gray
Green
Red
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Accuracy Color
21-369C 3 mL ±0.75% Green
22-146-316 10 mL ±0.6% Red
21-369E 5 mL ±0.75% Yellow
21-369D 4 mL ±0.75% White
21-369B 2 mL ±0.75% Red
21-369A 1 mL ±0.75% Gray
22-146-314 8 mL ±0.6% Red
22-146-315 9 mL ±0.6% Red
22-146-312 6 mL ±0.6% Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 21-369C Supplier VistaLab™ Technologies Supplier No. 1067
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

For large volume handling tasks such as reconstituting reagents, controls, or dilutions

  • Factory calibrated for accurate volume delivery
  • Can be easily adjusted to achieve accurate performance on dense or viscous liquids

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Accuracy ±0.75%
Autoclavable Not Autoclavable
Color Green
Disposable Reusable
For Use With Micropipette Tip
Material Aluminum, Stainless Steel
Model F1
Number of Channels 1
Sterility Non-sterile
Type Macropipette
Volume (Metric) 3 mL
Includes Nozzle Insert, Calibration Key
Quantity 1 Ea.
Warranty Lifetime
Length (English) 11 in.
Width (English) 2.5 in.
Height (English) 1.5 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.