Learn More
Aviva Systems Biology Corp MLC1 Antibody - middle region (ARP35249_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35249_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MLC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MLC1. Alternate names: KIAA0027, LVM, MLC, VLApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55681