Learn More
Enzo Life Sciences MLCK (342-352) amide (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP2090001
Encompass_Preferred
This sequence, KRRWKKNFIAV, is from skeletal muscle myosin light chain kinase (MLCK) (342-352) pseudosubstrate site. Inhibits MLCK (Ki =1 µM with MLC peptide substrate). Possesses only slight calmodulin binding activity. Alternative name: Myosin light chain kinase (342-352) amide, MLCK inhibitor. Purity: ≥97%. Long Term Storage: -20°C.
Catalog No. 50-201-1285