Learn More
Eurofins Discoverx MLCK, active, 250 µg. (14-638M)
Supplier:  Eurofins Discoverx 14638M
Encompass
N-terminal 6xHis-tagged recombinant, human MLCK amino acids 1425-1771. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9098