Learn More
Enzo Life Sciences MLCK substrate (skeletal and smooth muscle) (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP1140001
Encompass_Preferred
This sequence, KKRAARATSDVFA, is chicken gizzard smooth muscle myosin P-light chain (MLC 11-23) with Pro to Ala14 and Gln to Ala15 substitutions. Contains specificity determinants for MLC phosphorylation (target is Ser19) and is an excellent myosin light chain kinase (MLCK) substrate. Km=8.6 µM for rabbit skeletal muscle MLCK, 7.5 µM for chicken gizzard smooth muscle MLCK. Alternative name: Myosin light chain kinase substrate, Lys-Lys-Arg-Ala-Ala-Arg-Ala-Thr-Ser-Asn-Val-Phe-Ala-NH2. Purity: ≥97% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1233