Learn More
Aviva Systems Biology Corp MLF1 Antibody - N-terminal region (ARP53757_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53757_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MLF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MLF1. Alternate names: -ApplicationWB. Species reactivity: Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61118