Learn More
Aviva Systems Biology Corp MLF2 Antibody - middle region (ARP46600_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46600_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MLF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MLF2. Alternate names: NTN4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59358