Learn More
MilliporeSigma Milliflex Cassette prefilled with Sabouraud Dextrose Agar, 48/pk
Catalog No. NC0655273
Encompass
Medium designed for the recovery of a broad range of yeasts and molds found in various types of water, especially high purity water in the pharmaceutical industry.
Catalog No. NC0655273 Supplier MilliporeSigma Supplier No. MXSMCSD48
Medium designed for the recovery of a broad range of yeasts and molds found in various types of water, especially high purity water in the pharmaceutical industry.