Learn More
ATCC MLg; Lung ; Mouse (Mus musculus)
Supplier:  ATCC CCL206
Encompass
MLg [Mlg 2908] is a cell line exhibiting fibroblast morphology that was isolated in 1964 from the lung of a normal mouse (strain ddY). This cell line was deposited by Y Hirokawa.
Catalog No. 50-238-4507