Learn More
Aviva Systems Biology Corp MLH3 Antibody - middle region (ARP42403_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42403_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MLH3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MLH3. Alternate names: HNPCC7, MGC138372ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58214