Learn More
Apexbio Technology LLC MLi-2 1627091-47-7 2mg
SDP
Catalog No. 5000088776
Encompass_Preferred
MLi-2 (CAS 1627091-47-7) is an orally bioavailable highly selective inhibitor of LRRK2 (leucine-rich repeat kinase 2) a kinase implicated in the pathogenesis of Parkinson s disease MLi-2 demonstrates potent inhibitory activity with an IC50 of 0 76 nM By targeting LRRK2 MLi-2 is utilized in preclinical studies to investigate LRRK2-related signaling pathways and assess the therapeutic potential of LRRK2 inhibition in neurodegenerative disorders such as Parkinson s disease
Catalog No. 50-000-88776 Supplier Apexbio Technology LLC Supplier No. BA36832
MLi-2 (CAS 1627091-47-7) is an orally bioavailable highly selective inhibitor of LRRK2 (leucine-rich repeat kinase 2) a kinase implicated in the pathogenesis of Parkinson s disease MLi-2 demonstrates potent inhibitory activity with an IC50 of 0 76 nM By targeting LRRK2 MLi-2 is utilized in preclinical studies to investigate LRRK2-related signaling pathways and assess the therapeutic potential of LRRK2 inhibition in neurodegenerative disorders such as Parkinson s disease