Learn More
Medchemexpress LLC HY-100411 50mg , MLi-2 CAS:1627091-47-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10041150MG
Encompass_Preferred
HY-100411 50mg MLi-2 CAS: 1627091-47-7 MLi-2 is an orally active and highly selective LRRK2 inhibitor with an IC50 of 0.76 nM. MLi-2 has the potential for Parkinson’s disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0364