Learn More
Medchemexpress LLC MLK-IN-1 | 1627729-62-7 | 99.07% | 432.49 | 10 MG
SDP
Catalog No. 5000343739
Encompass_Preferred
MLK-IN-1 is a potent, brain penetrant and specific mixed lineage kinase 3 (MLK-3) inhibitor. It promotes continued axonogenesis in the presence of Tat-activated microglia and protects against effects of HIV-Tat in vitro.
  • Potent, brain penetrant, and specific mixed lineage kinase 3 (MLK-3) inhibitor.
  • Appearance: Solid, light yellow to green yellow.
  • For research use only.
  • Promotes continued axonogenesis in the presence of Tat-activated microglia in vitro.
  • Protects against the effects of HIV-Tat in vitro.
Catalog No. 50-003-43739 Supplier Medchemexpress LLC Supplier No. HY11135150MG
MLK-IN-1 is a potent, brain penetrant and specific mixed lineage kinase 3 (MLK-3) inhibitor. It promotes continued axonogenesis in the presence of Tat-activated microglia and protects against effects of HIV-Tat in vitro.
  • Potent, brain penetrant, and specific mixed lineage kinase 3 (MLK-3) inhibitor.
  • Appearance: Solid, light yellow to green yellow.
  • For research use only.
  • Promotes continued axonogenesis in the presence of Tat-activated microglia in vitro.
  • Protects against the effects of HIV-Tat in vitro.