Learn More
Medchemexpress LLC Mlk-in-1 | 1627729-62-7 | 99.1% | 432.49 g·mol⁻¹ | C23H20N4O3S | 5 MG
SDP
Catalog No. 5000264912
Encompass_Preferred
MLK-IN-1 is a research-grade small molecule that selectively inhibits mixed lineage kinase 3 (MLK-3). Described in patent US20140256733A1 as compound 68, it is reported to be potent and brain penetrant. The material is intended for laboratory research in both in vitro and in vivo neurobiological studies.
  • Selective MLK-3 inhibitor with reported potency.
  • Demonstrated brain penetration suitable for neurobiological studies.
  • High reported purity of 99.1%.
  • Molecular weight 432.49 g·mol⁻¹ and formula C23H20N4O3S.
  • Available in small quantities suitable for research use (for example, 5 mg).
Catalog No. 50-002-64912 Supplier Medchemexpress LLC Supplier No. HY1113515MG
MLK-IN-1 is a research-grade small molecule that selectively inhibits mixed lineage kinase 3 (MLK-3). Described in patent US20140256733A1 as compound 68, it is reported to be potent and brain penetrant. The material is intended for laboratory research in both in vitro and in vivo neurobiological studies.
  • Selective MLK-3 inhibitor with reported potency.
  • Demonstrated brain penetration suitable for neurobiological studies.
  • High reported purity of 99.1%.
  • Molecular weight 432.49 g·mol⁻¹ and formula C23H20N4O3S.
  • Available in small quantities suitable for research use (for example, 5 mg).