Learn More
Eurofins Discoverx MLK2, active, 10 µg. (14-964)
Supplier:  Eurofins Discoverx 14964
Encompass
N-terminal 6xHis-tagged recombinant, human MLK2 amino acids 1-449. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9496