Learn More
Carnabio Usa Inc MLK2(MAP3K10)/10UG/CATALYTIC
Catalog No. 507540878
Encompass
MLK2(MAP3K10), Catalytic domain, Wild type, Amino Acid 75-462, NP_002437.2, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0878 Supplier Carnabio Usa Inc Supplier No. 0911610
MLK2(MAP3K10), Catalytic domain, Wild type, Amino Acid 75-462, NP_002437.2, Expressed in Insect (sf21); 10microg