Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology MLL1 Monoclonal Antibody, Recombinant, Rabbit, 20 μL
SDP

Catalog No. 502808096
Encompass

MLL1 (E9W9Z) Rabbit Monoclonal Antibody (Carboxy-terminal Antigen) is a high-quality antibody designed to recognize endogenous levels of the total MLL1-C protein. This recombinant antibody is produced by immunizing animals with a recombinant protein specific to the carboxy terminus of the human MLL1 protein, a master regulator of hematopoiesis and embryogenesis. It is validated for use in various research applications, including western blotting, immunoprecipitation, and chromatin immunoprecipitation.

  • Recognizes endogenous levels of total MLL1-C protein
  • Validated for western blotting, immunoprecipitation, and chromatin immunoprecipitation
  • Host species: rabbit
  • Reactivity: human and monkey
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/ml BSA, 50% glycerol, and less than 0.02% sodium azide

Catalog No. 50-280-8096 Supplier Cell Signaling Technology Supplier No. 34907T
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MLL1 (E9W9Z) Rabbit Monoclonal Antibody (Carboxy-terminal Antigen) is a high-quality antibody designed to recognize endogenous levels of the total MLL1-C protein. This recombinant antibody is produced by immunizing animals with a recombinant protein specific to the carboxy terminus of the human MLL1 protein, a master regulator of hematopoiesis and embryogenesis. It is validated for use in various research applications, including western blotting, immunoprecipitation, and chromatin immunoprecipitation.

  • Recognizes endogenous levels of total MLL1-C protein
  • Validated for western blotting, immunoprecipitation, and chromatin immunoprecipitation
  • Host species: rabbit
  • Reactivity: human and monkey
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/ml BSA, 50% glycerol, and less than 0.02% sodium azide

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.