Learn More
Aviva Systems Biology Corp MLL4 Antibody - N-terminal region (ARP33704_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33704_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MLL4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MLL4. Alternate names: HRX2, MLL2, TRX2, WBP7, MLL4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-55120