Learn More
Aviva Systems Biology Corp AFDN Antibody - N-terminal region (ARP34225_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34225_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MLLT4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MLLT4. Alternate names: RP3-470B24.4, AF-6, AF6, AFADIN, FLJ34371, RP3-431P23.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-52946