Learn More
Aqua Solutions Molybdovanadate Reagent, 500 mL
SDP
Catalog No. NC1519075
Encompass
For phosphate (phosphorus) determination by the Molybdovanadate Method: Hach method 8114. Range: up to 45.0 mg/L PO4 3-. 500 mL bottle. Use with a bottle-top dispenser for Rapid Liquid procedures.
Catalog No. NC1519075 Supplier Aqua Solutions Supplier No. HA2076049
For phosphate (phosphorus) determination by the Molybdovanadate Method: Hach method 8114. Range: up to 45.0 mg/L PO4 3-. 500 mL bottle. Use with a bottle-top dispenser for Rapid Liquid procedures.