Learn More
Aviva Systems Biology Corp MLN Antibody - middle region (ARP44916_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44916_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MLN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MLN. Alternate names: MGC138519ApplicationWB. Species reactivity: Human, Sheep
Catalog No. 50-014-58935