Learn More
Medchemexpress LLC Mln120b dihydrochloride | 1782573-78-7 | 99.7% | 439.72 g·mol⁻¹ | C19H17Cl3N4O2 | 10 MG
SDP
Catalog No. 5000287451
Encompass_Preferred
MLN120B dihydrochloride is the dihydrochloride salt of MLN120B, a potent, ATP-competitive inhibitor of IKKβ used as a reference compound in NF-κB pathway and inflammatory disease research. It demonstrates an IC50 of approximately 60 nM against IKKβ and has been applied in biochemical, cellular, and in vivo studies. Supplied as a high-purity solid for laboratory research use only.
  • Potent ATP-competitive inhibitor of IKKβ (IC50 ≈ 60 nM).
  • Used in cellular and in vivo studies to inhibit multiple myeloma growth.
  • High purity (≈99.7%).
  • Suitable for biochemical and cellular assays.
  • Available in small vials (5 mg, 10 mg, 25 mg, 50 mg).
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-87451 Supplier Medchemexpress LLC Supplier No. HY15473A10MG
MLN120B dihydrochloride is the dihydrochloride salt of MLN120B, a potent, ATP-competitive inhibitor of IKKβ used as a reference compound in NF-κB pathway and inflammatory disease research. It demonstrates an IC50 of approximately 60 nM against IKKβ and has been applied in biochemical, cellular, and in vivo studies. Supplied as a high-purity solid for laboratory research use only.
  • Potent ATP-competitive inhibitor of IKKβ (IC50 ≈ 60 nM).
  • Used in cellular and in vivo studies to inhibit multiple myeloma growth.
  • High purity (≈99.7%).
  • Suitable for biochemical and cellular assays.
  • Available in small vials (5 mg, 10 mg, 25 mg, 50 mg).
  • Intended for research use only; not for human or veterinary use.