Learn More
Apexbio Technology LLC MLN2238(Synonyms: Ixazomib, MLN-2238, Ninlaro, MLN9708, Ixazomib citrate), 10mM (in 1mL DMSO), CAS: 1072833-77-2.
SDP
Catalog No. 5000060689
Encompass_Preferred
MLN2238 (CAS number 1072833-77-2) is a dipeptidyl boronic acid derivative designed as a reversible inhibitor of the 5 subunit (chymotrypsin-like activity) of the 20S proteasome with reported IC50 and Ki values of 3 4 nM and 0 93 nM respectively At higher concentrations MLN2238 also targets the proteasome s 1 (caspase-like) and 2 (trypsin-like) subunits In preclinical studies using xenograft and genetically-engineered models of hematologic malignancies MLN2238 exhibited potent antitumor activity promoting apoptosis and suppressing pathways such as NF- B MLN2238 has research applications in oncology particularly against multiple myeloma and lymphoma including in bortezomib-resistant cell lines
Catalog No. 50-000-60689 Supplier Apexbio Technology LLC Supplier No. A40085.1
 Restricted Item
Add to Cart
Add to Cart
MLN2238 (CAS number 1072833-77-2) is a dipeptidyl boronic acid derivative designed as a reversible inhibitor of the 5 subunit (chymotrypsin-like activity) of the 20S proteasome with reported IC50 and Ki values of 3 4 nM and 0 93 nM respectively At higher concentrations MLN2238 also targets the proteasome s 1 (caspase-like) and 2 (trypsin-like) subunits In preclinical studies using xenograft and genetically-engineered models of hematologic malignancies MLN2238 exhibited potent antitumor activity promoting apoptosis and suppressing pathways such as NF- B MLN2238 has research applications in oncology particularly against multiple myeloma and lymphoma including in bortezomib-resistant cell lines