Learn More
Medchemexpress LLC Pevonedistat | 905579-51-3 | 99.98% | C21H25N5O4S | 200 MG
SDP
Catalog No. 501639049
Encompass_Preferred
Pevonedistat (MLN4924) is a potent and selective NEDD8-activating enzyme (NAE) inhibitor. It acts by inducing deregulation of S-phase DNA synthesis, which disrupts protein turnover mediated by cullin-RING ligase, ultimately leading to apoptosis in human tumor cells. It also suppresses the growth of human tumor xenografts in mice.
  • Potent and selective NEDD8-activating enzyme (NAE) inhibitor
  • Induces deregulation of S-phase DNA synthesis
  • Disrupts protein turnover mediated by cullin-RING ligase
  • Promotes apoptosis of human tumor cells
  • Suppresses tumor xenograft growth in mice
Catalog No. 50-163-9049 Supplier Medchemexpress LLC Supplier No. HY70062200MG
Pevonedistat (MLN4924) is a potent and selective NEDD8-activating enzyme (NAE) inhibitor. It acts by inducing deregulation of S-phase DNA synthesis, which disrupts protein turnover mediated by cullin-RING ligase, ultimately leading to apoptosis in human tumor cells. It also suppresses the growth of human tumor xenografts in mice.
  • Potent and selective NEDD8-activating enzyme (NAE) inhibitor
  • Induces deregulation of S-phase DNA synthesis
  • Disrupts protein turnover mediated by cullin-RING ligase
  • Promotes apoptosis of human tumor cells
  • Suppresses tumor xenograft growth in mice