Learn More
Apexbio Technology LLC MLN4924 905579-51-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062171
Encompass_Preferred
MLN4924 (CAS 905579-51-3) is a selective inhibitor targeting NEDD8-activating enzyme (NAE) exhibiting an IC50 of 4 nM It acts competitively against AMP binding within the nucleotide-binding pocket of NAE with strong specificity over related enzymes UAE SAE UBA6 and ATG7 (IC50 1 5 M) In HCT-116 cells MLN4924 reduces Ubc12-NEDD8 and cullin-NEDD8 conjugation disrupting CRL-mediated ubiquitination and thus stabilizing substrates such as CDT1 resulting in cell cycle defects MLN4924 shows antitumor efficacy in HCT-116 H522 and Calu-6 tumor xenograft models highlighting its research utility in cancer biology
Catalog No. 50-000-62171 Supplier Apexbio Technology LLC Supplier No. B10365.1
MLN4924 (CAS 905579-51-3) is a selective inhibitor targeting NEDD8-activating enzyme (NAE) exhibiting an IC50 of 4 nM It acts competitively against AMP binding within the nucleotide-binding pocket of NAE with strong specificity over related enzymes UAE SAE UBA6 and ATG7 (IC50 1 5 M) In HCT-116 cells MLN4924 reduces Ubc12-NEDD8 and cullin-NEDD8 conjugation disrupting CRL-mediated ubiquitination and thus stabilizing substrates such as CDT1 resulting in cell cycle defects MLN4924 shows antitumor efficacy in HCT-116 H522 and Calu-6 tumor xenograft models highlighting its research utility in cancer biology