Learn More
Apexbio Technology LLC MLN4924, 50mg. Cas: 905579-51-3 MFCD: MFCD17215201. NAE inhibitor
SDP
Supplier:  Apexbio Technology LLC B103650
Encompass_Preferred
MLN4924 is a potent and selective inhibitor of NEDD8-activating enzyme (NAE) with IC50 value of 4nM [1].MLN4924 binds NAE within the nucleotide-binding site competing with AMP. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5200