Learn More
Medchemexpress LLC HY-10484 200mg , MLN4924 (hydrochloride) Pevonedistat hydrochloride CAS:1160295-21-5 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY10484200MG
Encompass_Preferred
Medchemexpress, HY-10484 200mg MLN4924 (hydrochloride) Pevonedistat hydrochloride CAS:1160295-21-5 Formula:C21H26ClN5O4S IC50: 4.7 nM (NAE) Purity:98% MLN4924 (hydrochloride) is a potent and selective NEDD8-activating enzyme ( NAE ) inhibitor, with IC 50 of 4.7 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2579