Learn More
Chemie Tek MLN7243, 1450833-55-2, MFCD28964160, 5 mg
SDP
Supplier:  Chemie Tek CTM72435MG
Encompass_Preferred
Formula: C19H20F3N5O5S2, MW: 519.5, Synonym: TAK-243, AOB 87172. MLN7243 (TAK-243) is a cell permeable small molecule inhibitor targeting ubiquitin-activating enzymes (UAE, also known as E1 enzymes).
Catalog No. NC1129905