Learn More
Selleck Chemical LLC MLN8054 5mg 869363-13-3
SDP
Catalog No. NC0398821
Encompass_Preferred
MLN8054 is a potent and selective inhibitor of Aurora A with IC50 of 4 nM in Sf9 insect cell. It is more than 40-fold selective for Aurora A than Aurora B. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0398821 Supplier Selleck Chemical LLC Supplier No. S11005MG
MLN8054 is a potent and selective inhibitor of Aurora A with IC50 of 4 nM in Sf9 insect cell. It is more than 40-fold selective for Aurora A than Aurora B. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.