Learn More
Apexbio Technology LLC MLN8237 (Alisertib), 1028486-01-2, MFCD16621243, 10 mg
SDP
Catalog No. 501012899
Encompass_Preferred
MF: C27H20ClFN4O4, MW: 518.92, Solubility: 25.9 mg/mL in DMSO,insoluble in ETOH, insoluble in H2O. The orally bioavailable agent, MLN8237 (also known as alisertib), is a potent small-molecule inhibitor of Aurora A kinase (AAK) which is overexpressed in several types of tumor and associated with oncogenesis and tumor progression.
Catalog No. 50-101-2899 Supplier Apexbio Technology LLC Supplier No. A411010
MF: C27H20ClFN4O4, MW: 518.92, Solubility: 25.9 mg/mL in DMSO,insoluble in ETOH, insoluble in H2O. The orally bioavailable agent, MLN8237 (also known as alisertib), is a potent small-molecule inhibitor of Aurora A kinase (AAK) which is overexpressed in several types of tumor and associated with oncogenesis and tumor progression.