Learn More
Apexbio Technology LLC MLN8237 (Alisertib) 1028486-01-2 200mg
SDP
Supplier:  Apexbio Technology LLC A4110200
Encompass_Preferred
MLN8237 (Alisertib CAS 1028486-01-2) is an orally administered small molecule inhibitor targeting Aurora A kinase (AAK) an enzyme frequently overexpressed in various cancers and linked to tumor proliferation and progression Developed from its predecessor MLN8054 to reduce benzodiazepine-like side effects MLN8237 potently inhibits Aurora A kinase through reversible ATP-competitive binding (Ki 0 43 nmol/L) Given its demonstrated anti-tumor activity in preclinical in vitro and in vivo studies MLN8237 is currently under investigation for treating advanced malignancies
Catalog No. 50-000-62533