Learn More
Apexbio Technology LLC MLN8237 (Alisertib), 50mg. Cas: 1028486-01-2 MFCD: MFCD16621243. Aurora A Kinase inhibitor, Potent and selective
SDP
Supplier:  Apexbio Technology LLC A411050
Encompass_Preferred
The orally bioavailable agent, MLN8237 (also known as alisertib), is a potent small-molecule inhibitor of Aurora A kinase (AAK) which is overexpressed in several types of tumor and associated with oncogenesis and tumor progression. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4883
 Restricted Item