Learn More
Selleck Chemical LLC Alisertib (MLN8237) 5mg 1028486-01-2
SDP
Supplier:  Selleck Chemical LLC S11335MG
Encompass_Preferred
Alisertib (MLN8237) is a selective Aurora A inhibitor with IC50 of 1.2 nM in a cell-free assay. It has >200-fold higher selectivity for Aurora A than Aurora B. Alisertib induces cell cycle arrest, apoptosis and autophagy. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-843-3