Learn More
Medchemexpress LLC HY-10452 100mg , MLN9708 Ixazomib citrate CAS:1239908-20-3 Purity:98%
SDP
Catalog No. 501493276
Encompass_Preferred
Medchemexpress, HY-10452 100mg MLN9708 Ixazomib citrate CAS:1239908-20-3 Formula:C20H23BCl2N2O9 IC50: 3.4 nM (20S proteasome β5), 31 nM (20S proteasome β1), 3500 nM (20S proteasome β2) Purity:98% MLN9708 rapidly hydrolyzes to MLN2238, which is a selective, orally bioavailable, second-generation proteasome inhibitor, inhibits the chymotrypsin-like proteolytic (β5) site of the 20S proteasome with an IC 50 value of 3.4 nM ( K i of 0.93 nM), and also inhibits the caspase-like (β1) and trypsin-like (β2) proteolytic sites with IC 50 of 31 and 3500 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3276 Supplier Medchemexpress LLC Supplier No. HY10452100MG
Medchemexpress, HY-10452 100mg MLN9708 Ixazomib citrate CAS:1239908-20-3 Formula:C20H23BCl2N2O9 IC50: 3.4 nM (20S proteasome β5), 31 nM (20S proteasome β1), 3500 nM (20S proteasome β2) Purity:98% MLN9708 rapidly hydrolyzes to MLN2238, which is a selective, orally bioavailable, second-generation proteasome inhibitor, inhibits the chymotrypsin-like proteolytic (β5) site of the 20S proteasome with an IC 50 value of 3.4 nM ( K i of 0.93 nM), and also inhibits the caspase-like (β1) and trypsin-like (β2) proteolytic sites with IC 50 of 31 and 3500 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.