Learn More
Aviva Systems Biology Corp MLPH Antibody - C-terminal region (ARP59494_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59494_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MLPH. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human MLPH. Alternate names: MGC2771, MGC59733, SLAC2-AApplicationWB. Species reactivity: Human
Catalog No. 50-286-720