Learn More
Selleck Chemical LLC MLR-1023-10mg
SDP
Catalog No. 502390013
Encompass_Preferred
MLR-1023 (Tolimidone, CP-26154) is a Lyn kinase activator and a novel insulin receptor potentiator that elicits a rapid-onset and durable improvement in glucose homeostasis in animal models of type 2 diabetes.
Catalog No. 50-239-0013 Supplier Selleck Chemical LLC Supplier No. S832110MG
MLR-1023 (Tolimidone, CP-26154) is a Lyn kinase activator and a novel insulin receptor potentiator that elicits a rapid-onset and durable improvement in glucose homeostasis in animal models of type 2 diabetes.