Learn More
TARGETMOL CHEMICALS INC MLS000544460 5MG
SDP
Catalog No. 502581486
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MLS000544460 is a highly selective and reversible Eya2 phosphatase inhibitor (Kd 2.0 uM IC50 4 uM).MLS000544460 exhibits inhibitory effects on Eya2 phosphatase-mediated cell migration and has anticancer activity. purity: 98%
Catalog No. 50-258-1486 Supplier TARGETMOL CHEMICALS INC Supplier No. T120805MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MLS000544460 is a highly selective and reversible Eya2 phosphatase inhibitor (Kd 2.0 uM IC50 4 uM).MLS000544460 exhibits inhibitory effects on Eya2 phosphatase-mediated cell migration and has anticancer activity. purity: 98%