Learn More
Medchemexpress LLC HY-112945 10mg Medchemexpress, MLS0315771 CAS:727664-91-7 Purity:>98%
SDP
Catalog No. 501969211
Encompass_Preferred
Medchemexpress, HY-112945 10mg MLS0315771 CAS:727664-91-7 MLS0315771 is a potent and biologically active competitive phosphomannose isomerase (MPI) inhibitor, with an IC50 ~1 μM and a Ki of 1.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9211 Supplier Medchemexpress LLC Supplier No. HY11294510MG
Medchemexpress, HY-112945 10mg MLS0315771 CAS:727664-91-7 MLS0315771 is a potent and biologically active competitive phosphomannose isomerase (MPI) inhibitor, with an IC50 ~1 μM and a Ki of 1.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.