Learn More
Aviva Systems Biology Corp MLSTD1 Antibody - C-terminal region (ARP47392_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47392_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MLSTD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human MLSTD1. Alternate names: FAR2, FLJ10462, MLSTD1, SDR10E2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59539