Learn More
Aviva Systems Biology Corp MLSTD2 Antibody - N-terminal region (ARP50044_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50044_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MLSTD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MLSTD2. Alternate names: DKFZp686A0370, DKFZp686P18247, FAR1, FLJ22728, FLJ33561, MLSTD2, SDR10E1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53614