Learn More
Medchemexpress LLC HY-115466 10mg Medchemexpress, MLT-748 CAS:1832578-30-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11546610MG
Encompass_Preferred
Medchemexpress, HY-115466 10mg MLT-748 CAS:1832578-30-9 MLT-748 is a potent, selective and allosteric inhibitor of MALT1, binds MALT1 in the allosteric Trp580 pocket, with an IC50 of 5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9384