Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MLT-748 10mM 1mL | 1832578-30-9 | 492.32 g/mol | C19H19Cl2N9O3 | 1 ML
SDP

Catalog No. 5000226724
Encompass_Preferred

MLT-748 is a potent, selective allosteric inhibitor of mucosa-associated lymphoid tissue lymphoma translocation protein 1 (MALT1). It binds the Trp580 allosteric pocket and inhibits MALT1 enzymatic activity with an IC50 of ≈5 nM. The compound is supplied as a 10 mM solution in DMSO (1 mL) and in solid forms for research applications.

  • Potent MALT1 inhibition (IC50 ≈ 5 nM).
  • Allosteric mechanism binding the Trp580 pocket.
  • High purity (≈99.9%).
  • Available as 10 mM solution in DMSO (1 mL) and as solids (5-100 mg).
  • Molecular formula C19H19Cl2N9O3; molecular weight ≈492.3 g/mol.
  • Suitable for biochemical and cellular assays probing MALT1 activity.

Catalog No. 50-002-26724 Supplier Medchemexpress LLC Supplier No. HY11546610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MLT-748 is a potent, selective allosteric inhibitor of mucosa-associated lymphoid tissue lymphoma translocation protein 1 (MALT1). It binds the Trp580 allosteric pocket and inhibits MALT1 enzymatic activity with an IC50 of ≈5 nM. The compound is supplied as a 10 mM solution in DMSO (1 mL) and in solid forms for research applications.

  • Potent MALT1 inhibition (IC50 ≈ 5 nM).
  • Allosteric mechanism binding the Trp580 pocket.
  • High purity (≈99.9%).
  • Available as 10 mM solution in DMSO (1 mL) and as solids (5-100 mg).
  • Molecular formula C19H19Cl2N9O3; molecular weight ≈492.3 g/mol.
  • Suitable for biochemical and cellular assays probing MALT1 activity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.