Learn More
Sarstedt Inc Cap, red, suitable for tubes Ø 10-16 mm, 1000/pack
Catalog No. NC1242350 Shop All Sarstedt Inc. Products
Encompass
Universal cap suitable for tubes with a diameter from 10 to 17 mm.
Catalog No. NC1242350 Supplier Sarstedt Inc Supplier No. 65.1379.002
Universal cap suitable for tubes with a diameter from 10 to 17 mm.