Learn More
MSC Dow Corning 99179259 10.1 oz Cartridge Gray RTV Silicone Joint Sealant -76 to 356°F Operating Temp, 20 min Tack Free Dry Time, 24 hr Full Cure Time, Series 732
Supplier:  MSC 00231720
Encompass
Dow Corning 99179259 10.1 oz Cartridge Gray RTV Silicone Joint Sealant -76 to 356°F Operating Temp, 20 min Tack Free Dry Time, 24 hr Full Cure Time, Series 732
Catalog No. 50-114-9224