Learn More
New England Biolabs, Inc. MluCI – 1000 units
SDP
Catalog No. 504458
Encompass
A restriction endonuclease that recognizes the sequence ^AATT_.
Catalog No. 50-445-8 Supplier New England Biolabs, Inc. Supplier No. R0538S
A restriction endonuclease that recognizes the sequence ^AATT_.