Learn More
New England Biolabs, Inc. MluCI – 5000 units
SDP
Catalog No. 504457
Encompass
A restriction endonuclease that recognizes the sequence ^AATT_.
Catalog No. 50-445-7 Supplier New England Biolabs, Inc. Supplier No. R0538L
A restriction endonuclease that recognizes the sequence ^AATT_.