Learn More
Aviva Systems Biology Corp MLX Antibody - N-terminal region (ARP39966_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39966_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MLX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MLX. Alternate names: MAD7, MXD7, TCFL4, bHLHd13ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57247