Learn More
Aviva Systems Biology Corp MLX Antibody - middle region (ARP51651_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51651_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MLX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MLX. Alternate names: MAD7, MXD7, TCFL4, bHLHd13ApplicationWB, IHC. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60576